Learn More
Medchemexpress LLC 2-Pyridinecarboxylic acid, 3-mercapto-, hydrochloride (1:1) | 320386-54-7 | 96.4% | 25 MG
SDP
Catalog No. 5000342398
Encompass_Preferred
SKF-34288 (3-Mercaptopicolinic acid) hydrochloride is an orally active phosphoenolpyruvate carboxykinase (PEPCK) inhibitor with a Ki of 2-9 μM. It functions as a potent hypoglycemic agent by inhibiting glucose synthesis, and also inhibits Asn metabolism, leading to an increase in amino acids and amides.
  • Orally active PEPCK inhibitor (Ki: 2-9 μM)
  • Potent hypoglycemic agent
  • Inhibits glucose synthesis
  • Inhibits Asn metabolism
  • Increases amino acids and amides
  • Inhibits C2C12 cell proliferation in vitro
  • Induces myogenic differentiation of C2C12 cells
  • Reduces mRNA expression of Pck2 and genes associated with serine biosynthesis
  • Reduces blood glucose in starved rats in vivo
  • Neutralizes Salbutamol-mediated hyperglycemia in rabbits
  • Reduces Pyruvate-induced blood glucose level in rats
Catalog No. 50-003-42398 Supplier Medchemexpress LLC Supplier No. 25MGHY128923
SKF-34288 (3-Mercaptopicolinic acid) hydrochloride is an orally active phosphoenolpyruvate carboxykinase (PEPCK) inhibitor with a Ki of 2-9 μM. It functions as a potent hypoglycemic agent by inhibiting glucose synthesis, and also inhibits Asn metabolism, leading to an increase in amino acids and amides.
  • Orally active PEPCK inhibitor (Ki: 2-9 μM)
  • Potent hypoglycemic agent
  • Inhibits glucose synthesis
  • Inhibits Asn metabolism
  • Increases amino acids and amides
  • Inhibits C2C12 cell proliferation in vitro
  • Induces myogenic differentiation of C2C12 cells
  • Reduces mRNA expression of Pck2 and genes associated with serine biosynthesis
  • Reduces blood glucose in starved rats in vivo
  • Neutralizes Salbutamol-mediated hyperglycemia in rabbits
  • Reduces Pyruvate-induced blood glucose level in rats