Learn More
Medchemexpress LLC 2-Pyridinecarboxylic acid, 5-hydroxy-, methyl ester | 30766-12-2 | 153.14 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00596310G
Encompass_Preferred
Methyl 5-hydroxypyridine-2-carboxylate is a phenolic acid that can be found in the stems of *Mahonia fortune*. It exhibits inhibitory effects on NO in LPS-stimulated RAW264.7 and BV2 cells and does not show cytotoxic effects at concentrations of 6.25-200 μM. This compound is soluble in DMSO and can be used in various *in vitro* and *in vivo* applications.
  • Found in the stems of *Mahonia fortune*
  • Inhibits NO in LPS-stimulated RAW264.7 and BV2 cells
  • Non-cytotoxic at concentrations of 6.25-200 μM
  • Soluble in DMSO
Catalog No. 50-004-48590