Learn More
Medchemexpress LLC 2-pyridinemethanol, α-[3-[(2R,6S)-2,6-dimethyl-1-piperidinyl]propyl]-α-phenyl hydrochloride | 61477-94-9 | MFCD01691662 | 98.8% | 374.95 g/mol | C22H31ClN2O | 50 MG
SDP
Catalog No. 5000213617
Encompass_Preferred
Pirmenol hydrochloride is the hydrochloride salt of pirmenol, an orally active antiarrhythmic compound used in cardiovascular research to study cardiac arrhythmias and ion channel modulation. It acts on muscarinic acetylcholine receptors to inhibit IK.ACh and is supplied as a solid for laboratory use.
  • Orally active antiarrhythmic compound used in research.
  • Inhibits IK.ACh by blocking muscarinic acetylcholine receptors (IC50 ~0.1 μM).
  • Provided as a white to yellow solid suitable for biochemical assays.
  • High reported purity (~98.8%) for analytical and pharmacology studies.
  • Recommended storage: solid at 4°C under inert gas; solutions at -20°C to -80°C.
Catalog No. 50-002-13617 Supplier Medchemexpress LLC Supplier No. HY100795A50MG
Pirmenol hydrochloride is the hydrochloride salt of pirmenol, an orally active antiarrhythmic compound used in cardiovascular research to study cardiac arrhythmias and ion channel modulation. It acts on muscarinic acetylcholine receptors to inhibit IK.ACh and is supplied as a solid for laboratory use.
  • Orally active antiarrhythmic compound used in research.
  • Inhibits IK.ACh by blocking muscarinic acetylcholine receptors (IC50 ~0.1 μM).
  • Provided as a white to yellow solid suitable for biochemical assays.
  • High reported purity (~98.8%) for analytical and pharmacology studies.
  • Recommended storage: solid at 4°C under inert gas; solutions at -20°C to -80°C.