Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-pyridinemethanol, α-[3-[(2R,6S)-2,6-dimethyl-1-piperidinyl]propyl]-α-phenyl-, hydrochloride | 61477-94-9 | MFCD01691662 | 99.3% | 374.95 g/mol | C22H31ClN2O | 1 ML
SDP

Catalog No. 5000213420
Encompass_Preferred

Pirmenol hydrochloride is an antiarrhythmic research compound supplied for laboratory use as a 10 mM solution in DMSO (1 mL) or as a solid with solvent. It inhibits IK.ACh and is intended for preclinical cardiovascular and ion-channel research. The material is characterized by high purity and is accompanied by a certificate of analysis.

  • Provided as 10 mM in DMSO, 1 mL stock solution or solid plus solvent
  • High purity, 99.3%
  • Molecular formula C22H31ClN2O; molecular weight 374.95 g/mol
  • CAS number 61477-94-9
  • Storage: solid at 4°C under nitrogen; in solvent -80°C for 6 months, -20°C for 1 month
  • Suitable for cardiovascular and ion-channel research applications

Catalog No. 50-002-13420 Supplier Medchemexpress LLC Supplier No. HY100795A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pirmenol hydrochloride is an antiarrhythmic research compound supplied for laboratory use as a 10 mM solution in DMSO (1 mL) or as a solid with solvent. It inhibits IK.ACh and is intended for preclinical cardiovascular and ion-channel research. The material is characterized by high purity and is accompanied by a certificate of analysis.

  • Provided as 10 mM in DMSO, 1 mL stock solution or solid plus solvent
  • High purity, 99.3%
  • Molecular formula C22H31ClN2O; molecular weight 374.95 g/mol
  • CAS number 61477-94-9
  • Storage: solid at 4°C under nitrogen; in solvent -80°C for 6 months, -20°C for 1 month
  • Suitable for cardiovascular and ion-channel research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.