Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Pyridinemethanol

Catalog No. P66602100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P66602100G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P66602100G Supplier Sigma Aldrich Supplier No. P66602100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(3,4-Dimethoxyphenyl)-1-propanol was used in the synthesis of fluorinated benzamide neuroleptic, (S)-N-[(1-ethyl-2- pyrrolidinyl)methyl]-5-(3-fluoropropyl)-2,3-dimethoxybenzamide.

Specifications

CAS 586-98-1
Density 1.131 g/mL (at 25°C (literature))
Boiling Point 112°C to 113°C (16 mmHg)
Molecular Formula C6H7NO
Refractive Index n20/D 1.543 (literature)
Linear Formula C6H7NO
MDL Number MFCD00006348
Quantity 5 g
Synonym omega-Hydroxy-2-picoline; 2-(Hydroxymethyl)pyridine; 2-Pyridyl carbinol
Molecular Weight (g/mol) 109.13
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.