Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridinemethanol 98.0+%, TCI America™
SDP

Catalog No. P0556100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0556100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0556100G Supplier TCI America Supplier No. P0556100G

missing translation for 'validMainframeMsg'

Specifications

CAS 586-98-1
Color Yellow
Boiling Point 112°C
Molecular Formula C6H7NO
MDL Number MFCD00006348
Quantity 100 g
Synonym 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol
InChI Key SHNUBALDGXWUJI-UHFFFAOYSA-N
SMILES C1=CC=NC(=C1)CO
IUPAC Name pyridin-2-ylmethanol
Molecular Weight (g/mol) 109.128
PubChem CID 11474
Formula Weight 109.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pyridinemethanol
Show More Show Less
EINECSNumber (5)-3694
RTECSNumber UT4689000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.