Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridinesulfonylacetonitrile, 98%

Catalog No. AAL0711406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0711406 5 g
AAL0711403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0711406 Supplier Thermo Scientific Chemicals Supplier No. L0711406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 170449-34-0 | C7H6N2O2S | 182.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170449-34-0
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.197
MDL Number MFCD00041443
InChI Key SAKACXZDAJXBCI-UHFFFAOYSA-N
Synonym 2-pyridinesulfonylacetonitrile, 2-pyridinesulphonylacetonitrile, 2-pyridin-2-ylsulfonyl acetonitrile, pyridine-2-sulfonylacetonitrile, 2-pyridine-2-sulfonyl acetonitrile, pyridin-2-ylsulfonyl acetonitrile, 2-2-pyridylsulfonyl ethanenitrile, maybridge1_004727, acmc-1c3jt, 2-pyridinesulfonlyacetonitrile
PubChem CID 519355
IUPAC Name 2-pyridin-2-ylsulfonylacetonitrile
SMILES C1=CC=NC(=C1)S(=O)(=O)CC#N

Specifications

Melting Point 77°C to 79°C
Quantity 5 g
UN Number UN3439
Formula Weight 182.21
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Pyridinesulfonylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement

H331-H302-H312

Toxic if inhaled.

Harmful if swallowed.

Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.