Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2-Pyridinylsulfanyl)acetic acid

Catalog No. CDS002508 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002508 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002508 Supplier Sigma Aldrich Supplier No. CDS00250850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The kinetic resolution of racemic 2-phenylbutyryl chloride by sterically hindered chiral secondary alcohols has been evaluated
  • 2-phenylbutyryl chloride reacts with 4-methoxybenzoyl chloride catalyzed by pdbr(pH)(pph3)2 to yield 1-(4-methoxyphenyl)-2-phenyl-2-buten-1-one.(r)-(-)-2-phenylbutyryl chloride may be employed as chiral reagent for the determination of dopamine and dopamine-derived salsolinol and norsalsolinol in human brain by GC-ms method
  • Chiral (s)-(+)-2-phenylbutyryl chloride may be used as derivatization reagent for the hydroxyl groups during the GC-ms assay for the enantiomers of 1,2-propanediol, 1,3-butanediol, 1,3-pentanediol and their corresponding hydroxyacids in biological fluids

Specifications

Molecular Formula C7H7NO2S
Linear Formula C7H7NO2S
MDL Number MFCD00276114
Quantity 5 mL
Molecular Weight (g/mol) 169.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.