Learn More
Medchemexpress LLC HY-101794 50mg Medchemexpress, 2-(Pyridyldithio)ethylamine (hydrochloride) CAS:106139-15-5 Purity:>98%
SDP
Catalog No. 501932220
Encompass_Preferred
Medchemexpress, HY-101794 50mg 2-(Pyridyldithio)ethylamine (hydrochloride) CAS:106139-15-5 2-(Pyridyldithio)ethylamine hydrochloride is a novel disulfide intercalating cross-linking reagent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2220 Supplier Medchemexpress LLC Supplier No. HY10179450MG
Medchemexpress, HY-101794 50mg 2-(Pyridyldithio)ethylamine (hydrochloride) CAS:106139-15-5 2-(Pyridyldithio)ethylamine hydrochloride is a novel disulfide intercalating cross-linking reagent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.