Learn More
Medchemexpress LLC HY-101794 500mg Medchemexpress, 2-(Pyridyldithio)ethylamine (hydrochloride) (S)-2-Pyridylthio Cysteamine Hydrochloride CAS:106139-15-5 Purity:98%
SDP
Catalog No. 501637016
Encompass_Preferred
Medchemexpress,HY-101794 500mg 2-(Pyridyldithio)ethylamine (hydrochloride) (S)-2-Pyridylthio Cysteamine Hydrochloride CAS:106139-15-5 Formula:C7H11ClN2S2 2-(Pyridyldithio)ethylamine hydrochloride is a novel disulfide intercalating cross-linking reagent. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7016 Supplier Medchemexpress LLC Supplier No. HY101794500MG
Medchemexpress,HY-101794 500mg 2-(Pyridyldithio)ethylamine (hydrochloride) (S)-2-Pyridylthio Cysteamine Hydrochloride CAS:106139-15-5 Formula:C7H11ClN2S2 2-(Pyridyldithio)ethylamine hydrochloride is a novel disulfide intercalating cross-linking reagent. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.