Learn More
Medchemexpress LLC (2-pyridyldithio)-PEG1-hydrazine | 111625-28-6 | MFCD22551638 | 99.3% | 187.28 g/mol | C7H9NOS2 | 250 MG
SDP
Catalog No. 5000203824
Encompass_Preferred
(2-pyridyldithio)-PEG1-hydrazine is a cleavable, single-unit PEG linker used for constructing antibody-drug conjugates (ADCs). It provides a pyridyldithio cleavable moiety and a hydrazine functional group for flexible conjugation and controlled payload release.
  • Cleavable pyridyldithio linker enabling disulfide-mediated payload release.
  • Single-unit PEG spacer to improve solubility and reduce steric hindrance.
  • Hydrazine functional handle for versatile conjugation chemistries.
  • Supplied in small research-scale quantities suitable for linker development.
  • Storage and stability guidance provided for refrigerated and solvent conditions.
Catalog No. 50-002-03824 Supplier Medchemexpress LLC Supplier No. HY136135250MG
(2-pyridyldithio)-PEG1-hydrazine is a cleavable, single-unit PEG linker used for constructing antibody-drug conjugates (ADCs). It provides a pyridyldithio cleavable moiety and a hydrazine functional group for flexible conjugation and controlled payload release.
  • Cleavable pyridyldithio linker enabling disulfide-mediated payload release.
  • Single-unit PEG spacer to improve solubility and reduce steric hindrance.
  • Hydrazine functional handle for versatile conjugation chemistries.
  • Supplied in small research-scale quantities suitable for linker development.
  • Storage and stability guidance provided for refrigerated and solvent conditions.