Learn More
Medchemexpress LLC (2-Pyridyldithio)-PEG2-Boc | 2144777-73-9 | >98% | C16H25NO4S2 | 250MG
SDP
Catalog No. 5000357340
Encompass_Preferred
(2-Pyridyldithio)-PEG2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds connect two ligands: one for an E3 ubiquitin ligase and another for a target protein. This design exploits the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based linker for PROTAC synthesis
  • Utilizes the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-57340 Supplier Medchemexpress LLC Supplier No. 250MGHY141361
(2-Pyridyldithio)-PEG2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds connect two ligands: one for an E3 ubiquitin ligase and another for a target protein. This design exploits the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based linker for PROTAC synthesis
  • Utilizes the ubiquitin-proteasome system
  • Enables selective degradation of target proteins