Learn More
Medchemexpress LLC (2-Pyridyldithio)-PEG4-propargyl | 2170240-99-8 | 98.5% | C16H23NO4S2 | 100 MG
SDP
Catalog No. 5000353114
Encompass_Preferred
(2-Pyridyldithio)-PEG4-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand with a target protein ligand.
  • For research use only
  • PROTAC linker
  • Suitable for click chemistry applications involving alkynes
  • High purity of 98.5%
Catalog No. 50-003-53114 Supplier Medchemexpress LLC Supplier No. HY141360100MG
(2-Pyridyldithio)-PEG4-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand with a target protein ligand.
  • For research use only
  • PROTAC linker
  • Suitable for click chemistry applications involving alkynes
  • High purity of 98.5%