Learn More
Medchemexpress LLC (2-Pyridyldithio)-PEG4-propargyl | 2170240-99-8 | 98.5% | C16H23NO4S2 | 50 MG
SDP
Catalog No. 5000353619
Encompass_Preferred
(2-Pyridyldithio)-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs connect two different ligands, one for an E3 ubiquitin ligase and the other for a target protein, to degrade target proteins using the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Connects two ligands for targeted protein degradation
  • Utilizes the intracellular ubiquitin-proteasome system
  • Appears as a colorless to light yellow liquid
  • Density of 1.18±0.1 g/cm3
Catalog No. 50-003-53619 Supplier Medchemexpress LLC Supplier No. HY14136050MG
(2-Pyridyldithio)-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs connect two different ligands, one for an E3 ubiquitin ligase and the other for a target protein, to degrade target proteins using the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Connects two ligands for targeted protein degradation
  • Utilizes the intracellular ubiquitin-proteasome system
  • Appears as a colorless to light yellow liquid
  • Density of 1.18±0.1 g/cm3