Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Pyrimidinecarboxylic acid, methyl ester | 34253-03-7 | 99.8% | C6H6N2O2 | 10 G
SDP

Catalog No. 5000435075
Encompass_Preferred

Methyl pyrimidine-2-carboxylate is a biochemical reagent suitable for life science research as a biological material or organic compound. It also acts as a synthetic intermediate in pharmaceutical synthesis.

  • Biochemical reagent for life science research
  • Useful as a synthetic intermediate for pharmaceutical synthesis
  • Appearance: Off-white to yellow solid
  • Purity: 99.76%
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year

Catalog No. 50-004-35075 Supplier Medchemexpress LLC Supplier No. HY7833710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Methyl pyrimidine-2-carboxylate is a biochemical reagent suitable for life science research as a biological material or organic compound. It also acts as a synthetic intermediate in pharmaceutical synthesis.

  • Biochemical reagent for life science research
  • Useful as a synthetic intermediate for pharmaceutical synthesis
  • Appearance: Off-white to yellow solid
  • Purity: 99.76%
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.