Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-pyrrolidin-1-ylaniline, Thermo Scientific™

Catalog No. CC44814DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC44814DE 5 g
CC44814CB 250 mg
CC44814DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC44814DE Supplier Thermo Scientific Chemicals Supplier No. CC44814DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21627-58-7
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
InChI Key BFPUBGCFJMIZDF-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl aniline, 2-pyrrolidinoaniline, 2-pyrrolidin-1-yl-phenylamine, 2-pyrrolidinylphenylamine, 2-pyrrolizinoaniline, acmc-209fmh, 2-pyrrolidin-1-ylphenylamine, 1-2-aminophenyl pyrrolidine, 2-pyrrolidin-1-ylphenyl amine, 2-1-pyrrolidinyl-benzenamine
PubChem CID 937501
IUPAC Name 2-pyrrolidin-1-ylaniline
SMILES C1CCN(C1)C2=CC=CC=C2N

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 2-Pyrrolidin-1-ylaniline
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.