Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Pyrrolidin-1-ylmethyl)quinazolin-4(3H)-one

Catalog No. OTV0005991G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0005991G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0005991G Supplier Sigma Aldrich Supplier No. OTV0005991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 6-chloro-1-hexyne is a halogenated hydrocarbon
  • It shows high reactivity in milling process during the synthesis of chloroalkyl-functionalized silicon nanoparticles due to the presence of triple bond.6-chloro-1-hexyne was used in the preparation of novel bis(indolyl)maleimide pyridinophanes and disubstituted polyacetylenes bearing naphthalene pendants with varying spacer lengths
  • It may be used in the preparation of diethyl 6-chloro-1-hexynylphosphonate, by lithiation with n-buli followed by the reaction with diethyl chlorophosphate

Specifications

Molecular Formula C13H15N3O
Linear Formula C13H15N3O
MDL Number MFCD00107510
Quantity 25 mL
Molecular Weight (g/mol) 229.28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.