Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Pyrrolidin-1-ylphenyl)methanol, 95%, Thermo Scientific™

Catalog No. CC44809CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC44809CB 250 mg Amber Glass Bottle
CC44809DA 1 g Amber Glass Bottle
CC44809DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC44809CB Supplier Thermo Scientific Chemicals Supplier No. CC44809CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73051-88-4
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
InChI Key QXIYEQQTAPJDBX-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-ylphenyl methanol, 2-pyrrolidin-1-yl phenyl methanol, 2-pyrrolizinobenzenemethanol, 2-1-pyrrolidinyl benzylalkohol, 2-pyrrolidinylphenyl methan-1-ol, benzenemethanol,2-1-pyrrolidinyl
PubChem CID 13021765
IUPAC Name (2-pyrrolidin-1-ylphenyl)methanol
SMILES C1CCN(C1)C2=CC=CC=C2CO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 177.25
Percent Purity 95%
Chemical Name or Material (2-Pyrrolidin-1-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.