Learn More
Medchemexpress LLC 2-Pyrrolidinecarboxamide, 1-[(2S)-2-(4-cyclopropyl-1H-1,2,3-triazol-1-yl)-3,3-dimethyl-1-oxobutyl]-4-hydroxy-N-[(5S)-6,7,8,9-tetrahydro-2-(4-methyl-5-thiazolyl)-5H-benzocyclohepten-5-yl]-, (2 S,4R)- | 2771069-76-0 | 99.19% | 576.75 | 25 MG
SDP
Catalog No. 5000395118
Encompass_Preferred
GNE7599 is a high-affinity and orally active von Hippel-Lindau (VHL) ligand with a Kd of 540 pM. It can be linked to a target protein ligand via a linker to form PROTACs.
  • High-affinity VHL ligand
  • Orally active
  • Can be connected to form PROTACs
Catalog No. 50-003-95118 Supplier Medchemexpress LLC Supplier No. 25MGHY162553
GNE7599 is a high-affinity and orally active von Hippel-Lindau (VHL) ligand with a Kd of 540 pM. It can be linked to a target protein ligand via a linker to form PROTACs.
  • High-affinity VHL ligand
  • Orally active
  • Can be connected to form PROTACs