Learn More
Medchemexpress LLC 2-Pyrrolidinepropanoic acid, 1-[(1,1-dimethylethoxy)carbonyl]-β-methoxy-α-methyl-, (αR,βR,2S)- | 120205-50-7 | 99.6% | 287.36 | 1 ML
SDP
Catalog No. 5000404058
Encompass_Preferred
N-Boc-dolaproine is the amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and has anticancer activity. N-Boc-dolaproine is a key intermediate used in the synthesis of Dolastatin 10.
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Has anticancer activity
  • Used as an intermediate in synthesis
  • Appearance: Colorless to light yellow oil
Catalog No. 50-004-04058 Supplier Medchemexpress LLC Supplier No. HY3304610MM/1ML
N-Boc-dolaproine is the amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and has anticancer activity. N-Boc-dolaproine is a key intermediate used in the synthesis of Dolastatin 10.
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Has anticancer activity
  • Used as an intermediate in synthesis
  • Appearance: Colorless to light yellow oil