Learn More
Medchemexpress LLC 2-Pyrrolidinepropanoic acid, 1-[(1,1-dimethylethoxy)carbonyl]-β-methoxy-α-methyl-, (αR,βR,2S)- | 1369427-40-6 | 99.9% | 468.67 | 250 MG
SDP
Catalog No. 5000402520
Encompass_Preferred
N-Boc-dolaproine dicyclohexylamine is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 is known for its ability to inhibit tubulin polymerization and mitosis, exhibiting anticancer activity. This product also contains dicyclohexylamine and is intended for research use only.
  • Amino acid residue of pentapeptide Dolastatin 10
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Exhibits anticancer activity
  • Contains dicyclohexylamine
  • For research use only
Catalog No. 50-004-02520 Supplier Medchemexpress LLC Supplier No. 250MGHY33046A
N-Boc-dolaproine dicyclohexylamine is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 is known for its ability to inhibit tubulin polymerization and mitosis, exhibiting anticancer activity. This product also contains dicyclohexylamine and is intended for research use only.
  • Amino acid residue of pentapeptide Dolastatin 10
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Exhibits anticancer activity
  • Contains dicyclohexylamine
  • For research use only