Learn More
Medchemexpress LLC 2-pyrrolidinone, 1-methyl-5-(3-pyridinyl-2,4,5,6-d4) | 350818-68-7 | 99.6% | 25 MG
SDP
Catalog No. 5000414521
Encompass_Preferred
(Rac)-Cotinine-d4 is the deuterium labeled (Rac)-Cotinine. It is for research use only and not sold to patients. This compound is used as a tracer and as an internal standard for quantitative analysis. Deuteration, which incorporates stable heavy isotopes into drug molecules, can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact the pharmacokinetic and metabolic profiles of pharmaceuticals
Catalog No. 50-004-14521 Supplier Medchemexpress LLC Supplier No. 25MGHYB1178S1
(Rac)-Cotinine-d4 is the deuterium labeled (Rac)-Cotinine. It is for research use only and not sold to patients. This compound is used as a tracer and as an internal standard for quantitative analysis. Deuteration, which incorporates stable heavy isotopes into drug molecules, can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact the pharmacokinetic and metabolic profiles of pharmaceuticals