Learn More
Medchemexpress LLC 2-Pyrrolidinone, 5-[(3S)-3-hydroxy-4-phenyl-1-buten-1-yl]-1-[6-(2H-tetrazol-5-yl)hexyl]- | 346673-06-1 | 98.3% | 383.49 | 50 MG
SDP
Catalog No. 5000346237
Encompass_Preferred
TCS 2510 is a selective EP4 agonist and can be used for the research of metabolic diseases. It stimulates glucagon-like peptide 1 and 2 secretion, activates the Gas signaling pathway, and increases Gcg mRNA expression levels. In vivo studies show it elevates plasma glucagon-like peptide 1 and 2 levels in mice.
  • Stimulates glucagon-like peptide 1 secretion.
  • Activates the Gas signaling pathway.
  • Increases Gcg mRNA expression levels.
  • Produces a concentration-dependent increase in cAMP levels in GLUTag cells.
  • Decreases cell impedance in GLUTag cells.
  • Produces a concentration-dependent increase in glucagon-like peptide 2 secretion.
  • Elevates plasma glucagon-like peptide 1 levels in fasted mice.
  • Increases plasma glucagon-like peptide 2 levels in mice.
Catalog No. 50-003-46237 Supplier Medchemexpress LLC Supplier No. HY108557500UG
TCS 2510 is a selective EP4 agonist and can be used for the research of metabolic diseases. It stimulates glucagon-like peptide 1 and 2 secretion, activates the Gas signaling pathway, and increases Gcg mRNA expression levels. In vivo studies show it elevates plasma glucagon-like peptide 1 and 2 levels in mice.
  • Stimulates glucagon-like peptide 1 secretion.
  • Activates the Gas signaling pathway.
  • Increases Gcg mRNA expression levels.
  • Produces a concentration-dependent increase in cAMP levels in GLUTag cells.
  • Decreases cell impedance in GLUTag cells.
  • Produces a concentration-dependent increase in glucagon-like peptide 2 secretion.
  • Elevates plasma glucagon-like peptide 1 levels in fasted mice.
  • Increases plasma glucagon-like peptide 2 levels in mice.