Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-Pyrrolidinone-5-carboxylic acid, 99%

Catalog No. AAL0942430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0942430 250 g
AAL0942418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0942430 Supplier Thermo Scientific Chemicals Supplier No. L0942430
Add to Cart

CAS: 149-87-1 | C5H7NO3 | 129.12 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (almost transparency)

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-87-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00064322
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid
PubChem CID 499
ChEBI CHEBI:16010
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Melting Point 183°C to 185°C
Quantity 250 g
Beilstein 82131
Solubility Information Soluble in water. (almost transparency)
Formula Weight 129.12
Percent Purity 99%
Chemical Name or Material (+/-)-2-Pyrrolidinone-5-carboxylic acid
Hazard Category H318
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 205-748-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.