Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyrrolidinone, 99%

Catalog No. AC132110025
Change view
Click to view available options
Quantity:
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132110025 2.5 L Glass bottle
AC132112500 250 mL Glass bottle
AC132115000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC132110025 Supplier Thermo Scientific Chemicals Supplier No. 132110025
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 616-45-5 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-45-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00005270
InChI Key HNJBEVLQSNELDL-UHFFFAOYSA-N
Synonym 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
PubChem CID 12025
ChEBI CHEBI:36592
IUPAC Name pyrrolidin-2-one
SMILES C1CC(=O)NC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 25.5°C
Color Colorless to Yellow
Density 1.1030g/mL
Boiling Point 250.0°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4860 to 1.4880
Quantity 2.5 L
Beilstein 21, 236
Specific Gravity 1.103
Merck Index 15, 8130
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ethanol, ether, chloroform, benzene,, ethyl acetate, carbon disulfide
Viscosity 10.2 mPa.s (30°C)
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid or Low Melting Mass
Chemical Name or Material 2-Pyrrolidinone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
EINECSNumber 210-483-1
RTECSNumber UY5715000
TSCA TSCA
Recommended Storage Product may darken during storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.