Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Quinolinecarbonitrile, 97%

Catalog No. AC449580050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449580050 5 g Glass bottle
AC449580010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449580050 Supplier Thermo Scientific Chemicals Supplier No. 449580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1436-43-7 | C10H6N2 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1436-43-7
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00134341
InChI Key WDXARTMCIRVMAE-UHFFFAOYSA-N
Synonym 2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0
PubChem CID 74031
IUPAC Name quinoline-2-carbonitrile
SMILES N#CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 93°C to 95°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 154.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Quinolinecarbonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H StatementHarmful if swallowed.Harmful in contact with skin.Harmful if inhaled.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation.
Precautionary Statement GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsing.
EINECSNumber 215-865-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.