Learn More
Medchemexpress LLC 2-quinolinecarboxaldehyde, 8-hydroxy- | 14510-06-6 | MFCD00168962 | 98.0% | 173.17 g/mol | C10H7NO2 | 1 G
SDP
Catalog No. 5000301016
Encompass_Preferred
8-Hydroxyquinoline-2-carbaldehyde is an organic biochemical reagent (C10H7NO2, MW 173.17 g/mol) used as a building block in organic synthesis and as a ligand in coordination and analytical chemistry. It is supplied as a solid powder in laboratory pack sizes for research applications, with batch-specific certificates of analysis available for quality confirmation.
  • Acts as a building block for organic synthesis.
  • Functions as a bidentate ligand and chelating agent.
  • Suitable for analytical and proteomics applications.
  • Provided in small pack sizes appropriate for laboratory research.
  • Batch-specific certificate of analysis available for purity verification.
Catalog No. 50-003-01016 Supplier Medchemexpress LLC Supplier No. HYW0168401G
8-Hydroxyquinoline-2-carbaldehyde is an organic biochemical reagent (C10H7NO2, MW 173.17 g/mol) used as a building block in organic synthesis and as a ligand in coordination and analytical chemistry. It is supplied as a solid powder in laboratory pack sizes for research applications, with batch-specific certificates of analysis available for quality confirmation.
  • Acts as a building block for organic synthesis.
  • Functions as a bidentate ligand and chelating agent.
  • Suitable for analytical and proteomics applications.
  • Provided in small pack sizes appropriate for laboratory research.
  • Batch-specific certificate of analysis available for purity verification.