Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-quinolinecarboxaldehyde, 8-hydroxy- | 14510-06-6 | MFCD00168962 | 99.6% | 173.17 g·mol⁻1 | C10H7NO2 | 250 MG
SDP

Catalog No. 5000302585
Encompass_Preferred

8-Hydroxyquinoline-2-carbaldehyde is an organic biochemical reagent used as a synthetic intermediate, analytical standard, and research reagent in life-science and chemical synthesis applications. It contains a hydroxyl-substituted quinoline aldehyde core and is supplied in small laboratory pack sizes for experimental use.

  • High purity suitable for analytical and synthetic work (99.6% by HPLC).
  • Molecular formula C10H7NO2 and molecular weight 173.17 g·mol⁻1.
  • Light yellow to orange solid appearance.
  • Available in small lab pack sizes for research use.
  • Store at -20°C, sealed, away from moisture.
  • Includes batch certificate of analysis with identity and purity data.

Catalog No. 50-003-02585 Supplier Medchemexpress LLC Supplier No. 250MGHYW016840
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Hydroxyquinoline-2-carbaldehyde is an organic biochemical reagent used as a synthetic intermediate, analytical standard, and research reagent in life-science and chemical synthesis applications. It contains a hydroxyl-substituted quinoline aldehyde core and is supplied in small laboratory pack sizes for experimental use.

  • High purity suitable for analytical and synthetic work (99.6% by HPLC).
  • Molecular formula C10H7NO2 and molecular weight 173.17 g·mol⁻1.
  • Light yellow to orange solid appearance.
  • Available in small lab pack sizes for research use.
  • Store at -20°C, sealed, away from moisture.
  • Includes batch certificate of analysis with identity and purity data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.