Learn More
Medchemexpress LLC 2-quinolinecarboxylic acid, 4-hydroxy-, sodium salt (1:1) | 196901 | MFCD12195841 | 99.8% | 211.15 g/mol | C10H6NNaO3 | 1 ML
SDP
Catalog No. 5000240658
Encompass_Preferred
Kynurenic acid sodium is the sodium salt of kynurenic acid, an endogenous tryptophan metabolite that functions as a broad-spectrum antagonist of ionotropic glutamate receptors and α7 nicotinic acetylcholine receptors. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and also offered as a solid form, it is used in vitro for receptor pharmacology, electrophysiology, and biochemical assays.
  • Ready-to-use 10 mM solution in DMSO, 1 mL
  • Also available as solid form for custom preparation
  • Reported purity 99.8%
  • Suitable for receptor pharmacology and electrophysiology assays
  • CAS number 2439-02-3 for unambiguous identification
Catalog No. 50-002-40658 Supplier Medchemexpress LLC Supplier No. HY10751210MM 1ML
Kynurenic acid sodium is the sodium salt of kynurenic acid, an endogenous tryptophan metabolite that functions as a broad-spectrum antagonist of ionotropic glutamate receptors and α7 nicotinic acetylcholine receptors. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and also offered as a solid form, it is used in vitro for receptor pharmacology, electrophysiology, and biochemical assays.
  • Ready-to-use 10 mM solution in DMSO, 1 mL
  • Also available as solid form for custom preparation
  • Reported purity 99.8%
  • Suitable for receptor pharmacology and electrophysiology assays
  • CAS number 2439-02-3 for unambiguous identification