Learn More
Medchemexpress LLC 2-Quinoxalinecarboxamide, N-[2-[3-[(4-amino-1-piperidinyl)methyl]imidazo[2,1-b]thiazol-6-yl]phenyl]- | 925437-38-3 | 96.2% | 483.59 | 500 UG
SDP
Catalog No. 5000385638
Encompass_Preferred
Sirtuin modulator 8 is a sirtuin modulator that can activate Sirtuin activity. It significantly downregulates the expression of monocyte chemoattractant protein-1 (MCP-1) and upregulates the expression of fibroblast growth factor 21 (FGF21). This modulator regulates the expression of inflammation- and metabolism-related genes, showing potential for studying metabolic diseases and inflammatory diseases. It also causes a significant decrease in MCP-1 expression in a human leukocyte (WBC) model, indicating its potential to inhibit inflammatory responses.
  • Activates sirtuin activity.
  • Downregulates MCP-1 expression.
  • Upregulates FGF21 expression.
  • Shows potential for studying metabolic diseases.
  • Shows potential for studying inflammatory diseases.
Catalog No. 50-003-85638 Supplier Medchemexpress LLC Supplier No. HY173463500UG
Sirtuin modulator 8 is a sirtuin modulator that can activate Sirtuin activity. It significantly downregulates the expression of monocyte chemoattractant protein-1 (MCP-1) and upregulates the expression of fibroblast growth factor 21 (FGF21). This modulator regulates the expression of inflammation- and metabolism-related genes, showing potential for studying metabolic diseases and inflammatory diseases. It also causes a significant decrease in MCP-1 expression in a human leukocyte (WBC) model, indicating its potential to inhibit inflammatory responses.
  • Activates sirtuin activity.
  • Downregulates MCP-1 expression.
  • Upregulates FGF21 expression.
  • Shows potential for studying metabolic diseases.
  • Shows potential for studying inflammatory diseases.