Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2'R)-2'-Deoxy-2'-fluoro-2'-methyluridine 98.0+%, TCI America™
SDP

Catalog No. D4823200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4823200MG 200 mg
D48231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4823200MG Supplier TCI America Supplier No. D4823200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 863329-66-2
Molecular Formula C10H13FN2O5
Molecular Weight (g/mol) 260.22
MDL Number MFCD19982668
InChI Key ARKKGZQTGXJVKW-UHFFFAOYNA-N
Synonym PSI-6206
PubChem CID 11311503
IUPAC Name 1-[3-fluoro-4-hydroxy-5-(hydroxymethyl)-3-methyloxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1(F)C(O)C(CO)OC1N1C=CC(=O)NC1=O

Specifications

Melting Point 234°C
Color White
Quantity 200 mg
Formula Weight 260.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2′R)-2′-Deoxy-2′-fluoro-2′-methyluridine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.