Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(S)-1-Hydroxyethyl]pyridine 97.0+%, TCI America™
SDP

Catalog No. H12631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12631G Supplier TCI America Supplier No. H12631G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59042-90-9
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD06795465
InChI Key PPHIIIRFJKDTLG-UHFFFAOYNA-N
Synonym s-1-pyridin-2-yl ethanol, s-1-2-pyridyl ethanol, 2-s-1-hydroxyethyl pyridine, s---2-1-hydroxyethyl pyridine, s-2-1-hydroxyethyl pyridine, 1s-1-pyridin-2-yl ethan-1-ol, 1s-1-pyridin-2-yl ethanol, s-1-2-pyridyl ethanol, s-2-1-hydroxyethyl pyridine, pubchem5701, 1s-1-pyridin-2-ylethanol
PubChem CID 11094597
IUPAC Name 1-(pyridin-2-yl)ethan-1-ol
SMILES CC(O)C1=CC=CC=N1

Specifications

Melting Point 29°C
Color White-Yellow
Quantity 1 g
Formula Weight 123.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 2-[(S)-1-Hydroxyethyl]pyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.