Learn More
Medchemexpress LLC 2-(S)-(3,5-bis(4-(trifluoromethyl)phenyl)phenyl)-4-methylpentanoic acid | 1146594-87-7 | 99.5% | 480.44 | C26H22F6O2 | 1 MG
SDP
Catalog No. 5000214773
Encompass_Preferred
JNJ-40418677 is a small-molecule γ-secretase modulator used in preclinical Alzheimer's disease research. It is orally active and brain-penetrant, and has been shown to selectively lower amyloid β42 production and reduce plaque burden in animal models, while sparing Notch processing.
Catalog No. 50-002-14773 Supplier Medchemexpress LLC Supplier No. HY1006041MG
JNJ-40418677 is a small-molecule γ-secretase modulator used in preclinical Alzheimer's disease research. It is orally active and brain-penetrant, and has been shown to selectively lower amyloid β42 production and reduce plaque burden in animal models, while sparing Notch processing.