Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-sec-butyl-3-methoxypyrazine | 24168-70-5 | MFCD00006136 | 99.0% | 166.22 | C9H14N2O | 1 G
SDP

Catalog No. 5000301822
Encompass_Preferred

2-Sec-butyl-3-methoxypyrazine (SBMP) is a methoxypyrazine compound used in analytical and sensory research into odor-active molecules, flavor and fragrance chemistry, and chemical ecology. It is naturally occurring in some insects and is supplied as a high-purity liquid reagent for laboratory use.

  • High purity suitable for analytical and research applications.
  • Molecular formula C9H14N2O and molecular weight 166.22.
  • Available as a liquid, with solution formats in DMSO and solid pack sizes for flexibility.
  • Commonly used in flavor, fragrance, and chemical ecology studies.
  • Density approximately 1 g/cm3, facilitating volumetric handling.

Catalog No. 50-003-01822 Supplier Medchemexpress LLC Supplier No. HYW0171401G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Sec-butyl-3-methoxypyrazine (SBMP) is a methoxypyrazine compound used in analytical and sensory research into odor-active molecules, flavor and fragrance chemistry, and chemical ecology. It is naturally occurring in some insects and is supplied as a high-purity liquid reagent for laboratory use.

  • High purity suitable for analytical and research applications.
  • Molecular formula C9H14N2O and molecular weight 166.22.
  • Available as a liquid, with solution formats in DMSO and solid pack sizes for flexibility.
  • Commonly used in flavor, fragrance, and chemical ecology studies.
  • Density approximately 1 g/cm3, facilitating volumetric handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.