Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-sec-butyl-3-methoxypyrazine (SBMP) | 24168-70-5 | MFCD00006136 | 99.0% | 166.22 | C9H14N2O | 5 G
SDP

Catalog No. 5000299861
Encompass_Preferred

2-Sec-butyl-3-methoxypyrazine (SBMP) is a methoxypyrazine provided as an analytical standard for use in flavor, fragrance, and natural-product research. It is intended for research use and is commonly used in analytical workflows for identification and quantification of methoxypyrazines in biological and food samples.

  • CAS number: 24168-70-5.
  • Molecular formula C9H14N2O; molecular weight 166.22 g/mol.
  • High purity suitable for analytical applications (99.04%).
  • Appearance: colorless to light yellow liquid; density ~1 g/cm3.
  • Storage: pure form -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Available package sizes include 1 g, 5 g, 10 g, 25 g, and 50 g.

Catalog No. 50-002-99861 Supplier Medchemexpress LLC Supplier No. HYW0171405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Sec-butyl-3-methoxypyrazine (SBMP) is a methoxypyrazine provided as an analytical standard for use in flavor, fragrance, and natural-product research. It is intended for research use and is commonly used in analytical workflows for identification and quantification of methoxypyrazines in biological and food samples.

  • CAS number: 24168-70-5.
  • Molecular formula C9H14N2O; molecular weight 166.22 g/mol.
  • High purity suitable for analytical applications (99.04%).
  • Appearance: colorless to light yellow liquid; density ~1 g/cm3.
  • Storage: pure form -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Available package sizes include 1 g, 5 g, 10 g, 25 g, and 50 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.