Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-sec-Butylthiazole, 98+%

Catalog No. AAA1348314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1348314 25 g
AAA1348306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1348314 Supplier Thermo Scientific Chemicals Supplier No. A1348314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18277-27-5 | C7H11NS | 141.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18277-27-5
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.23
MDL Number MFCD00051951
InChI Key MHJSWOZJMPIGJQ-UHFFFAOYNA-N
Synonym 2-sec-butylthiazole, 2-sec-butyl thiazole, 2-1-methylpropyl thiazole, 2-1-methylpropyl-thiazole, thiazole, 2-1-methylpropyl, 2-butan-2-yl-1,3-thiazole, thiazole, 2-sec-butyl, fema no. 3372, 2-s-butylthiazole
PubChem CID 519539
IUPAC Name 2-butan-2-yl-1,3-thiazole
SMILES CCC(C)C1=NC=CS1

Specifications

Density 1.007
Boiling Point 174°C to 175°C
Flash Point 58°C (136°F)
Odor Characteristic
Refractive Index 1.4985
Quantity 25 g
UN Number UN1993
Formula Weight 141.24
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-sec-Butylthiazole
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement

H226

Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 242-154-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.