Learn More
Medchemexpress LLC 2-selenouracil | 16724-03-1 | 98.0% | 175.05 g·mol⁻¹ | C4H4N2OSe | 5 MG
SDP
Catalog No. 5000264112
Encompass_Preferred
2-Selenouracil is a small-molecule photosensitizer used in photodynamic therapy research. It produces reactive oxygen species upon light activation and is supplied for laboratory research use only.
  • Useful specialized photosensitizer for photodynamic therapy research.
  • Generates reactive oxygen species (ROS) under light activation.
  • Chemical formula C4H4N2OSe; molecular weight 175.05 g·mol⁻¹.
  • High purity, typically 98.0% as supplied.
  • Available in small laboratory quantities, such as 5 mg, for research applications.
  • Accompanied by a product datasheet describing handling and specifications.
Catalog No. 50-002-64112 Supplier Medchemexpress LLC Supplier No. HY1119595MG
2-Selenouracil is a small-molecule photosensitizer used in photodynamic therapy research. It produces reactive oxygen species upon light activation and is supplied for laboratory research use only.
  • Useful specialized photosensitizer for photodynamic therapy research.
  • Generates reactive oxygen species (ROS) under light activation.
  • Chemical formula C4H4N2OSe; molecular weight 175.05 g·mol⁻¹.
  • High purity, typically 98.0% as supplied.
  • Available in small laboratory quantities, such as 5 mg, for research applications.
  • Accompanied by a product datasheet describing handling and specifications.