Learn More
Chemglass Life Sciences Vacuum Pump, Welch, Belt Driven, 90L/min, 0.1 micron Ultimate Vacuum, Motor: 1/2 Hp and 525 RPM, 2.1L Oil Capacity, 7/16" ID Of Tubing Required, 112lbs, 20" L x 12" W x 15" H, MUST SHIP TRUCK
SDP
Supplier:  Chemglass Life Sciences AF035001
Encompass_Preferred
Welch general purpose 2 stage belt driven pumps designed to operate at lower RPM allowing them to run cooler and reach an ultimate vacuum of 1 x 10-4 Torr. Lower speed operation also reduces the wear on the pump components ensuring a longer life. All pumps are equipped with a gas ballast. Pumps operate on 115V, 60Hz and can be ordered for 230V operation.Long Service Life due to Low Pump RPM Suited for:¿ Freeze Drying¿ Concentrating¿ Schlenk Line¿ Degassing¿ Vacuum Distillations¿ Vacuum DryingVacuum Pump, Welch, Belt Driven, 90L/min, 0.1 micron Ultimate Vacuum, Motor: 1/2 Hp and 525 RPM, 2.1L Oil Capacity, 7/16" ID Of Tubing Required, 112lbs, 20" L x 12" W x 15" H, MUST SHIP TRUCK
Catalog No. AF035001