Learn More
Diversified Spaces™ 2-Station Service Center with Sink, Single Faced
GSA_VASCI_ED
Catalog No. S38144 Shop All Diversified Spaces Products
Change view
Click to view available options
Top Style:
0.75 in. Phenolic Resin Surface
1 in. Solid Epoxy Resin Surface
Includes:
1 sink, 2 Cold Water/Gas Fixtures, a GFI-protected AC duplex receptacle, 2 Rod Sockets, 4 Locking Drawers and Locking Cupboard with Shelf
1 sink, 2 Cold Water/Gas Fixtures, a GFI-protected AC duplex receptacle, 2 Rod Sockets, Locking Cupboard
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Top Style Includes
S38144 0.75 in. Phenolic Resin Surface 1 sink, 2 Cold Water/Gas Fixtures, a GFI-protected AC duplex receptacle, 2 Rod Sockets, 4 Locking Drawers and Locking Cupboard with Shelf
S38146 1 in. Solid Epoxy Resin Surface 1 sink, 2 Cold Water/Gas Fixtures, a GFI-protected AC duplex receptacle, 2 Rod Sockets, 4 Locking Drawers and Locking Cupboard with Shelf
S38178 1 in. Solid Epoxy Resin Surface 1 sink, 2 Cold Water/Gas Fixtures, a GFI-protected AC duplex receptacle, 2 Rod Sockets, Locking Cupboard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S38144 Supplier Diversified Spaces™ Supplier No. C2514K
Only null left
Add to Cart
Add to Cart
Single-faced table that provides a fully equipped science center for two students. Diversified Spaces™™ 2-Station Service Center has a solid epoxy resin sink along with cold water faucets for every two students.
  • Two student single faced workstation includes a sink, two cold water/gas fixtures, a GFI protected AC duplex receptacle and two rod sockets
  • Includes two book compartments in the aprons
  • Constructed of solid oak and oak hardwood veneers
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Limited Lifetime Warranty
  • Made in the U.S.A.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Lab Table
Autoclavable Not Autoclavable
Color Oak
No. of Shelves None
Includes 1 sink, 2 Cold Water/Gas Fixtures, a GFI-protected AC duplex receptacle, 2 Rod Sockets, 4 Locking Drawers and Locking Cupboard with Shelf
Top Style 0.75 in. Phenolic Resin Surface
With Tilt No
Footrest No
No. of Doors 1
No. of Drawers 4
Voltage 110 V
With Casters No
Plug Type GFI protected AC duplex receptacle
Certifications/Compliance SEFA compliant and MAS Certified Green
Dimensions (D x W x H) 30 x 66 x 36 in.
Finish Chemical Resistant, Earth-friendly UV
Total Positions 2
Material Oak with Hardwood Veneer