Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S 2-Stop Precision Pump Tubing, Tygon E-LFL, L/S 16; Pack of 8
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0644716/DEL
Encompass
Applications: General laboratory use Characteristics: Long life, won¿t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent For use with Masterflex multichannel pump heads.
Catalog No. NC0870299