Learn More
Precision Glassblowing of Colorado 2 STOP PVC RED-RED TUBNG 1.143
SDP
Catalog No. NC9374959
Encompass_Preferred
2-Stop Red/Red (0.044 in./1.14mm ID), PVC Pump Tubing, 12/Pack
Catalog No. NC9374959 Supplier Precision Glassblowing of Colorado Supplier No. PT2180P
2-Stop Red/Red (0.044 in./1.14mm ID), PVC Pump Tubing, 12/Pack