Learn More
AVANTOR MASTERFLEX (VWR LLC) Tubing, 2-Stop, E-Lab, 0.44mm ID, 12/pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9646016/DEL
Encompass
Ismatec SC0004-LT 2-Stop, E-Lab Tubing, 0.44mm ID, 12/pk, Designed for optimal operation in your Ismatec pumpsLong service life - outstanding fatigue resistance due to flexing!Application: Suitable for applications using acetone, methyl-ethyl-ketone (MEK), and other highly aggressive chemicalsResistant to ozone and UV light ¿ won't crack, swell, or hardenTranslucent yellow colorTemp range: -50 to 74¿C (-58 to 165¿F)
Catalog No. NC1128996