Learn More
Elemental Scientific Inc 2 STP PVC TBNG .27MM 18IN PK12
SDP
Catalog No. NC0567608
Encompass_Preferred
Peristaltic Pump Tubing, PVC, 2-stop, 0.27mm ID, 2.0mm flared 151mm bridge distance (orange/blue), 12 pc.
Catalog No. NC0567608 Supplier Elemental Scientific Inc Supplier No. ES43971027
Peristaltic Pump Tubing, PVC, 2-stop, 0.27mm ID, 2.0mm flared 151mm bridge distance (orange/blue), 12 pc.