Learn More
Elemental Scientific Inc 2 STP PVC TBNG .64MM 18IN PK12
SDP
Catalog No. NC0567607
Encompass_Preferred
Peristaltic Pump Tubing, PVC, 2-stop, 0.64mm ID, 2.0mm flared 151mm bridge distance (orange/white), 12 pc.
Catalog No. NC0567607 Supplier Elemental Scientific Inc Supplier No. ES43971064
Peristaltic Pump Tubing, PVC, 2-stop, 0.64mm ID, 2.0mm flared 151mm bridge distance (orange/white), 12 pc.