Learn More
AVANTOR MASTERFLEX (VWR LLC) 2-STP TYGN E-LAB 0.51MMID 12PK
Catalog No. 502741934
Encompass
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-Lab, 0.51 mm ID; 12/PK, Avantor
Catalog No. 50-274-1934 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9646018
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-Lab, 0.51 mm ID; 12/PK, Avantor