Learn More
AVANTOR MASTERFLEX (VWR LLC) 2-STP TYGN E-LAB 2.54MMID 12PK
Catalog No. 502741916
Encompass
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-Lab, 2.54 mm ID; 12/PK, Avantor
Catalog No. 50-274-1916 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9646046
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-Lab, 2.54 mm ID; 12/PK, Avantor