Learn More
AVANTOR MASTERFLEX (VWR LLC) 2-STP TYGN E-LFL 2.20MMID 12PK
Catalog No. 502741966
Encompass
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-LFL, 2.20 mm ID; 12/PK, Avantor
Catalog No. 50-274-1966 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9644943
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-LFL, 2.20 mm ID; 12/PK, Avantor