Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(tert-Butoxycarbonylamino)phenylboronic acid, 97%

Catalog No. AC445340010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445340010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445340010 Supplier Thermo Scientific Chemicals Supplier No. 445340010

missing translation for 'validMainframeMsg'

CAS: 146140-95-6 | C11H16BNO3 | 221.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146140-95-6
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD01114645
InChI Key MXRAJVMTCAUABO-UHFFFAOYSA-N
Synonym 2-tert-butylcarbonylamino phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivaloylamino phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivaloylaminobenzene boronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl
PubChem CID 4193502
IUPAC Name [2-(2,2-dimethylpropanoylamino)phenyl]boronic acid
SMILES CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O

Specifications

Melting Point 296.5°C to 297.5°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 221.06
Percent Purity 97%
Chemical Name or Material 2-(tert-Butoxycarbonylamino)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.