Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-tert-Butyl-1,1,3,3-tetramethylguanidine

Catalog No. 206155ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-615-5ML 5 mL
20-615-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-615-5ML Supplier Sigma Aldrich Supplier No. 206155ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-tert-Butyl-1,1,3,3-tetramethylguanidine (Bartonαs base) is an excellent alternative to traditional inorganic bases for promoting the coupling reaction.

Specifications

CAS 29166-72-1
Boiling Point 88°C to 89°C (43 mmHg)
Molecular Formula C9H21N3
Linear Formula C9H21N3
Refractive Index n20/D 1.457
Quantity 5 mL
MDL Number MFCD01862972
Synonym BTMG
Molecular Weight (g/mol) 171.28
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.