Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-tert-butyl-1,4-benzoquinone | 3602-55-9 | 99.0% | 164.20 g·mol-1 | C10H12O2 | 100 G
SDP

Catalog No. 5000301826
Encompass_Preferred

2-tert-butyl-1,4-benzoquinone is an electrophilic benzoquinone used as a research reagent in biochemical and toxicology studies. It is reported as a metabolite of butylated hydroxyanisole and an oxidation product of 2-tert-butylhydroquinone; supplied as a light-yellow solid at high purity.

  • High purity (99.0%).
  • Molecular formula C10H12O2; molecular weight 164.20 g·mol-1.
  • Light-yellow to yellow solid appearance.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Used in biochemical and toxicology research, including studies of metabolism and apoptosis.

Catalog No. 50-003-01826 Supplier Medchemexpress LLC Supplier No. HYW017187100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-tert-butyl-1,4-benzoquinone is an electrophilic benzoquinone used as a research reagent in biochemical and toxicology studies. It is reported as a metabolite of butylated hydroxyanisole and an oxidation product of 2-tert-butylhydroquinone; supplied as a light-yellow solid at high purity.

  • High purity (99.0%).
  • Molecular formula C10H12O2; molecular weight 164.20 g·mol-1.
  • Light-yellow to yellow solid appearance.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Used in biochemical and toxicology research, including studies of metabolism and apoptosis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.